Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Nitrophenyl)ethylamine hydrochloride, ≥98%

Click to view available options
Quantity:
1g
5g
25g
29968-78-3
C8H11ClN2O2
202.638
MFCD00012900
JVMHULJEYUQYSH-UHFFFAOYSA-N
4-nitrophenethylamine hydrochloride,4-nitrophenylethylamine hydrochloride,4-nitrophenethylamine hcl,2-4-nitrophenyl ethanamine hydrochloride,2-4-nitrophenyl ethylamine hydrochloride,p-nitrophenylethylamine hcl,benzeneethanamine, 4-nitro-, monohydrochloride,4-nitrophenethylaminehydrochloride,p-nitrophenethylamine, hydrochloride,pubchem11627
458983
2-(4-nitrophenyl)ethanamine;hydrochloride
C1=CC(=CC=C1CCN)[N+](=O)[O-].Cl
29968-78-3
C8H11ClN2O2
202.638
MFCD00012900
JVMHULJEYUQYSH-UHFFFAOYSA-N
4-nitrophenethylamine hydrochloride,4-nitrophenylethylamine hydrochloride,4-nitrophenethylamine hcl,2-4-nitrophenyl ethanamine hydrochloride,2-4-nitrophenyl ethylamine hydrochloride,p-nitrophenylethylamine hcl,benzeneethanamine, 4-nitro-, monohydrochloride,4-nitrophenethylaminehydrochloride,p-nitrophenethylamine, hydrochloride,pubchem11627
458983
2-(4-nitrophenyl)ethanamine;hydrochloride
C1=CC(=CC=C1CCN)[N+](=O)[O-].Cl
Applications
4-Nitrophenethylamine hydrochloride was used in the synthesis of ortho-metalated primary phenethylamines complexes containing six-membered palladacycles and N-(4-nitrophenethyl)formamide.

Solubility
Soluble in methanol almost transparency.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 1g
Melting Point 200°C (literature)
Color Undesignated
Beilstein 3568915
Formula Weight 202.64
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material 2-(4-Nitrophenyl)ethylamine hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning