Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Pentanedione, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
500g
1kg
2.5kg
10kg
123-54-6
C5H8O2
100.117
YRKCREAYFQTBPV-UHFFFAOYSA-N
2,4-pentanedione,acetylacetone,2,4-pentadione,diacetylmethane,acetoacetone,acac,2,4-dioxopentane,pentanedione,pentan-2,4-dione,acetyl acetone
31261
CHEBI:14750
pentane-2,4-dione
CC(=O)CC(=O)C
123-54-6
C5H8O2
100.117
YRKCREAYFQTBPV-UHFFFAOYSA-N
2,4-pentanedione,acetylacetone,2,4-pentadione,diacetylmethane,acetoacetone,acac,2,4-dioxopentane,pentanedione,pentan-2,4-dione,acetyl acetone
31261
CHEBI:14750
pentane-2,4-dione
CC(=O)CC(=O)C
Color Undesignated
Density 0.9750g/mL
Quantity 500g
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Water 0.1% max.
Infrared Spectrum Authentic
Specific Gravity 97%2 to 97%8 (20°C)
Assay Percent Range 99+%
Linear Formula CH3COCH2COCH3
Formula Weight 100.12
Physical Form Liquid
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2, 4-Pentanedione
Show More Show Less