Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Pentanedione dioxime, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
2157-56-4
C5H10N2O2
130.147
MFCD00013932
WBRYLZHYOFBTPD-YDFGWWAZSA-N
acetylacetone dioxime,2,4-pentanedione dioxime,2,4-pentanedione, dioxime,2,4-pentanedioxime,2,4-pentanedione oxime,2,4-pentanedione,2,4-dioxime,2,4-pentanedione, 2,4-dioxime,pentane-2,4-dione oxime,2e,4e-2,4-pentanedione dioxime #,ne-n-4e-4-hydroxyiminopentan-2-ylidene hydroxylamine
9573501
(NE)-N-[(4E)-4-hydroxyiminopentan-2-ylidene]hydroxylamine
CC(=NO)CC(=NO)C
2157-56-4
C5H10N2O2
130.147
MFCD00013932
WBRYLZHYOFBTPD-YDFGWWAZSA-N
acetylacetone dioxime,2,4-pentanedione dioxime,2,4-pentanedione, dioxime,2,4-pentanedioxime,2,4-pentanedione oxime,2,4-pentanedione,2,4-dioxime,2,4-pentanedione, 2,4-dioxime,pentane-2,4-dione oxime,2e,4e-2,4-pentanedione dioxime #,ne-n-4e-4-hydroxyiminopentan-2-ylidene hydroxylamine
9573501
(NE)-N-[(4E)-4-hydroxyiminopentan-2-ylidene]hydroxylamine
CC(=NO)CC(=NO)C
Melting Point 145°C to 149°C
Packaging Glass bottle
Quantity 5g
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 130.15
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 4-Pentanedione dioxime, 98%
Show More Show Less

EINECSNumber : 218-472-3

TSCA : TSCA