Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Pentanedione, Reagent, 99%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
5 gal.
Packaging:
Amber Glass Bottle
Steel Can
123-54-6
C5H8O2
100.12
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
CC(=O)CC(C)=O
123-54-6
C5H8O2
100.12
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
CC(=O)CC(C)=O

P1016, 123-54-6, C5H8O2

Spectrum™ 2,4-Pentanedione, Reagent , also known as acetylacetone, is also a building block for the synthesis of heterocyclic compounds. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Packaging Amber Glass Bottle
Quantity 500 mL
CAS Max % 1
UN Number UN2310
Percent Purity 99%