Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Pyridinedicarboxylic Acid +99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
499-80-9
C7H5NO4·xH2O
167.12
MFCD00149413
MJIVRKPEXXHNJT-UHFFFAOYSA-N
2,4-pyridinedicarboxylic acid,lutidinic acid,2,4-lutidinic acid,2,4-pyridinedicarboxylicacid, hydrate 1:1,4nrq,pd2,4-carboxypicolinic acid,pubchem16314,pyridine carboxylate, 6a,acmc-1bnb9
10365
CHEBI:44737
pyridine-2,4-dicarboxylic acid
C1=CN=C(C=C1C(=O)O)C(=O)O
499-80-9
C7H5NO4·xH2O
167.12
MFCD00149413
MJIVRKPEXXHNJT-UHFFFAOYSA-N
2,4-pyridinedicarboxylic acid,lutidinic acid,2,4-lutidinic acid,2,4-pyridinedicarboxylicacid, hydrate 1:1,4nrq,pd2,4-carboxypicolinic acid,pubchem16314,pyridine carboxylate, 6a,acmc-1bnb9
10365
CHEBI:44737
pyridine-2,4-dicarboxylic acid
C1=CN=C(C=C1C(=O)O)C(=O)O
Melting Point 243°C to 246°C
Color White
Water 10% max. (K.F.)
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula (C5H3N)(COOH)2·xH2O
Beilstein 22, 153
Solubility Information Solubility in water: insoluble
Formula Weight 167.12
Physical Form Crystalline Powder
Percent Purity ≥99.0% (on dry substance)
Chemical Name or Material 2, 4-Pyridinedicarboxylic acid hydrate, 99+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 207-892-2

TSCA : TSCA