Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Pyridyl)benzimidazole 98.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
2208-59-5
C12H9N3
195.23
MFCD00453948
UYWWLYCGNNCLKE-UHFFFAOYSA-N
2-4-pyridyl benzimidazole, 2-pyridin-4-yl-1h-benzo d imidazole, 2-pyridin-4-yl-1h-benzimidazole, 2-pyridin-4-yl-1h-1,3-benzodiazole, 2-4-pyridyl-1h-benzimidazole, 2-pyridin-4-yl-1h-benzoimidazole, 2-benzimidazole, acmc-1ch3h, cambridge id 5231114, 2-4-pyridinyl-benzimidazole
247634
2-(pyridin-4-yl)-1H-1,3-benzodiazole
N1C2=CC=CC=C2N=C1C1=CC=NC=C1
2208-59-5
C12H9N3
195.23
MFCD00453948
UYWWLYCGNNCLKE-UHFFFAOYSA-N
2-4-pyridyl benzimidazole, 2-pyridin-4-yl-1h-benzo d imidazole, 2-pyridin-4-yl-1h-benzimidazole, 2-pyridin-4-yl-1h-1,3-benzodiazole, 2-4-pyridyl-1h-benzimidazole, 2-pyridin-4-yl-1h-benzoimidazole, 2-benzimidazole, acmc-1ch3h, cambridge id 5231114, 2-4-pyridinyl-benzimidazole
247634
2-(pyridin-4-yl)-1H-1,3-benzodiazole
N1C2=CC=CC=C2N=C1C1=CC=NC=C1
Color Red-Yellow
Melting Point 218°C
Quantity 5g
Formula Weight 195.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material 2-(4-Pyridyl)benzimidazole