Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Quinolinediol, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
86-95-3
C9H7NO2
161.16
HDHQZCHIXUUSMK-UHFFFAOYSA-N
2,4-quinolinediol,2,4-dihydroxyquinoline,4-hydroxyquinolin-2 1h-one,quinoline-2,4-diol,4-hydroxy-2-quinolone,4-hydroxycarbostyril,2 1h-quinolinone, 4-hydroxy,4-hydroxy-2-quinolinone,hydroxycarbostyril,2-hydroxyquinolin-4 1h-one
54680871
CHEBI:75926
4-hydroxy-1H-quinolin-2-one
C1=CC=C2C(=C1)C(=CC(=O)N2)O
86-95-3
C9H7NO2
161.16
HDHQZCHIXUUSMK-UHFFFAOYSA-N
2,4-quinolinediol,2,4-dihydroxyquinoline,4-hydroxyquinolin-2 1h-one,quinoline-2,4-diol,4-hydroxy-2-quinolone,4-hydroxycarbostyril,2 1h-quinolinone, 4-hydroxy,4-hydroxy-2-quinolinone,hydroxycarbostyril,2-hydroxyquinolin-4 1h-one
54680871
CHEBI:75926
4-hydroxy-1H-quinolin-2-one
C1=CC=C2C(=C1)C(=CC(=O)N2)O
Quantity 50g
Color Brown
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 21, 171
Formula Weight 161.16
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2, 4-Quinolinediol
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning