Alfa Aesar™ 2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H2705114

Quantity 25g

 View more versions of this product

Catalog No. AAH2705114

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C18H29BO2
    Formula Weight 288.24
    MDL Number MFCD09953477
    Synonym 2-[2-(4-tert-Butylphenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Chemical Name or Material 2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester

    Specifications

    Formula Weight 288.24
    Molecular Formula C18H29BO2
    MDL Number MFCD09953477
    Synonym 2-[2-(4-tert-Butylphenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Quantity 25g
    Chemical Name or Material 2-(4-tert-Butylphenyl)ethylboronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit