Alfa Aesar™ 2,5,6-Trimethylbenzothiazole, 98%

Manufacturer: Alfa Aesar™ B2184606

Quantity 5g

 View more versions of this product

Catalog No. AAB2184606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5683-41-0
    Molecular Formula C10H11NS
    Formula Weight 177.26
    MDL Number MFCD00128832
    Chemical Name or Material 2,5,6-Trimethylbenzothiazole

    Specifications

    Formula Weight 177.26
    Molecular Formula C10H11NS
    MDL Number MFCD00128832
    CAS 5683-41-0
    Odor Odorless
    Boiling Point 143°C (5mmHg)
    Melting Point 83° to 85°C
    Quantity 5g
    Chemical Name or Material 2,5,6-Trimethylbenzothiazole
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit