Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5,6-Trimethylbenzothiazole, 98%

Click to view available options
Quantity:
1g
5g
25g
5683-41-0
C10H11NS
177.265
MFCD00128832
ZZLZXZFVVOXVRG-UHFFFAOYSA-N
2,5,6-trimethylbenzothiazole,2,5,6-trimethylbenzo d thiazole,benzothiazole, 2,5,6-trimethyl,pubchem20514,acmc-1b075,benzothiazole,2,5,6-trimethyl,zzlzxzfvvoxvrg-uhfffaoysa,benzothiazole, 2,5,6-trimethyl-6ci,7ci,8ci,9ci
79765
2,5,6-trimethyl-1,3-benzothiazole
CC1=CC2=C(C=C1C)SC(=N2)C
5683-41-0
C10H11NS
177.265
MFCD00128832
ZZLZXZFVVOXVRG-UHFFFAOYSA-N
2,5,6-trimethylbenzothiazole,2,5,6-trimethylbenzo d thiazole,benzothiazole, 2,5,6-trimethyl,pubchem20514,acmc-1b075,benzothiazole,2,5,6-trimethyl,zzlzxzfvvoxvrg-uhfffaoysa,benzothiazole, 2,5,6-trimethyl-6ci,7ci,8ci,9ci
79765
2,5,6-trimethyl-1,3-benzothiazole
CC1=CC2=C(C=C1C)SC(=N2)C
Quantity 25g
Odor Odorless
Melting Point 83°C to 85°C
Boiling Point 143°C (5mmHg)
Formula Weight 177.26
Percent Purity 98%
Chemical Name or Material 2,5,6-Trimethylbenzothiazole

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 227-148-0

TSCA : No

Recommended Storage : Ambient temperatures