Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5,6-Trimethylbenzoxazole, 98%

Click to view available options
Quantity:
1g
5g
25g
19219-98-8
C10H11NO
161.204
MFCD00022849
PDVDINPEWGXOHX-UHFFFAOYSA-N
2,5,6-trimethylbenzoxazole,benzoxazole, 2,5,6-trimethyl,2,5,6-trimethylbenzo d oxazole,pubchem8696,2,6-trimethylbenzoxazole,acmc-1c5qd,2,5,6-trimethylbenzoxazol,benzoxazole,5,6-trimethyl,2,5,6-trimethyl-benzoxazole,benzoxazole,2,5,6-trimethyl
87965
2,5,6-trimethyl-1,3-benzoxazole
CC1=CC2=C(C=C1C)OC(=N2)C
19219-98-8
C10H11NO
161.204
MFCD00022849
PDVDINPEWGXOHX-UHFFFAOYSA-N
2,5,6-trimethylbenzoxazole,benzoxazole, 2,5,6-trimethyl,2,5,6-trimethylbenzo d oxazole,pubchem8696,2,6-trimethylbenzoxazole,acmc-1c5qd,2,5,6-trimethylbenzoxazol,benzoxazole,5,6-trimethyl,2,5,6-trimethyl-benzoxazole,benzoxazole,2,5,6-trimethyl
87965
2,5,6-trimethyl-1,3-benzoxazole
CC1=CC2=C(C=C1C)OC(=N2)C
Quantity 1g
Melting Point 91°C to 96°C
Formula Weight 161.2
Percent Purity 98%
Chemical Name or Material 2,5,6-Trimethylbenzoxazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 242-887-9

TSCA : Yes

Recommended Storage : Ambient temperatures