Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5,6-Trimethylbenzoxazole, 98%

Click to view available options
Quantity:
1g
5g
25g
19219-98-8
C10H11NO
161.204
MFCD00022849
PDVDINPEWGXOHX-UHFFFAOYSA-N
2,5,6-trimethylbenzoxazole,benzoxazole, 2,5,6-trimethyl,2,5,6-trimethylbenzo d oxazole,pubchem8696,2,6-trimethylbenzoxazole,acmc-1c5qd,2,5,6-trimethylbenzoxazol,benzoxazole,5,6-trimethyl,2,5,6-trimethyl-benzoxazole,benzoxazole,2,5,6-trimethyl
87965
2,5,6-trimethyl-1,3-benzoxazole
CC1=CC2=C(C=C1C)OC(=N2)C
19219-98-8
C10H11NO
161.204
MFCD00022849
PDVDINPEWGXOHX-UHFFFAOYSA-N
2,5,6-trimethylbenzoxazole,benzoxazole, 2,5,6-trimethyl,2,5,6-trimethylbenzo d oxazole,pubchem8696,2,6-trimethylbenzoxazole,acmc-1c5qd,2,5,6-trimethylbenzoxazol,benzoxazole,5,6-trimethyl,2,5,6-trimethyl-benzoxazole,benzoxazole,2,5,6-trimethyl
87965
2,5,6-trimethyl-1,3-benzoxazole
CC1=CC2=C(C=C1C)OC(=N2)C
Quantity 5g
Melting Point 91°C to 96°C
Formula Weight 161.2
Percent Purity 98%
Chemical Name or Material 2,5,6-Trimethylbenzoxazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA