Alfa Aesar™ 2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole, 98%

Manufacturer: Alfa Aesar™ H5104103

Quantity 1g

 View more versions of this product

Catalog No. AAH5104103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 146370-52-7
    Molecular Formula C17H26Cl2O2
    Formula Weight 333.29
    MDL Number MFCD03093939
    UN Number UN1759
    Chemical Name or Material 2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole

    Specifications

    Formula Weight 333.29
    Molecular Formula C17H26Cl2O2
    MDL Number MFCD03093939
    CAS 146370-52-7
    UN Number UN1759
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Chemical Name or Material 2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole
    Assay 98%

    Description

    Applications
    2,5-Bis(chloromethyl)-4-(2-ethylhexyloxy)anisole is used for conducting polymer precursor. It is also used as organic chemical synthesis intermediate.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit