Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-7028799
Click to view available options
Quantity:
1 g
395-64-2
C9H4F6O
242.12
MFCD00674087
PICGJIQKPLBIES-UHFFFAOYSA-N
2,5-bis trifluoromethyl benzaldehyde, benzaldehyde, 2,5-bis trifluoromethyl, pubchem10107, acmc-1ajin, attercop-chm at107097, 2,5-bistrifluoromethyl benzaldehyde, 2,5-bis-trifluoromethyl-benzaldehyde
2737807
2,5-bis(trifluoromethyl)benzaldehyde
FC(F)(F)C1=CC=C(C(C=O)=C1)C(F)(F)F
395-64-2
C9H4F6O
242.12
MFCD00674087
PICGJIQKPLBIES-UHFFFAOYSA-N
2,5-bis trifluoromethyl benzaldehyde, benzaldehyde, 2,5-bis trifluoromethyl, pubchem10107, acmc-1ajin, attercop-chm at107097, 2,5-bistrifluoromethyl benzaldehyde, 2,5-bis-trifluoromethyl-benzaldehyde
2737807
2,5-bis(trifluoromethyl)benzaldehyde
FC(F)(F)C1=CC=C(C(C=O)=C1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.468
Boiling Point 137°C
Flash Point 69°C (157°F)
Refractive Index 1.422
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 242.12
Percent Purity 97%
Chemical Name or Material 2,5-Bis(trifluoromethyl)benzaldehyde
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit