Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Bis(trifluoromethyl)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H2659103

Quantity 1g

 View more versions of this product

Catalog No. AAH2659103


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

51012-27-2
239.12
UN3276
C9H3F6N
MFCD03094423
2,5-Bis(trifluoromethyl)benzonitrile

Specifications

51012-27-2
239.12
UN3276
2,5-Bis(trifluoromethyl)benzonitrile
C9H3F6N
MFCD03094423
1g
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H227-H312-H332-H315-H319
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit