Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Bis(trifluoromethyl)bromobenzene, 97%, Acros Organics

Click to view available options
Quantity:
5g
7617-93-8
C8H3BrF6
293.006
GFQNSGHVOFVTLC-UHFFFAOYSA-N
2,5-bis trifluoromethyl bromobenzene,2-bromo-1,4-bis trifluoromethyl benzene,1-bromo-2,5-bis trifluoromethyl benzene,1,4-bis trifluoromethyl-2-bromobenzene,2,5-bis trifluoromethyl bromobenzene,benzene, 2-bromo-1,4-bis trifluoromethyl,pubchem4264,acmc-1bisb,ksc495k7j,2,5-bis-trifluoromethylbromobenzene
2734763
2-bromo-1,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)C(F)(F)F
7617-93-8
C8H3BrF6
293.006
GFQNSGHVOFVTLC-UHFFFAOYSA-N
2,5-bis trifluoromethyl bromobenzene,2-bromo-1,4-bis trifluoromethyl benzene,1-bromo-2,5-bis trifluoromethyl benzene,1,4-bis trifluoromethyl-2-bromobenzene,2,5-bis trifluoromethyl bromobenzene,benzene, 2-bromo-1,4-bis trifluoromethyl,pubchem4264,acmc-1bisb,ksc495k7j,2,5-bis-trifluoromethylbromobenzene
2734763
2-bromo-1,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)C(F)(F)F
Refractive Index 1.4325 to 1.4345 (20°C, 589nm)
Quantity 5g
Color Colorless
Infrared Spectrum Authentic
Density 1.691g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CF3)C6H3Br
Formula Weight 293.01
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2, 5-Bis(trifluoromethyl)bromobenzene, 97%
Show More Show Less