Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,5-Di-tert-butyl-4-methoxyphenol, 97%
Click to view available options
Quantity:
5g
25g
Description
Applications
2,5-Di-tert-butyl-4-methoxyphenol is used as a non-tumorigenic compound in contrast to DBHQ.
Notes
Incompatible with strong oxidizing agents.
2,5-Di-tert-butyl-4-methoxyphenol is used as a non-tumorigenic compound in contrast to DBHQ.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 99°C to 102°C |
Color | White to Cream |
Quantity | 5g |
Formula Weight | 236.36 |
Physical Form | Crystalline Powder |
Percent Purity | 97% |
Chemical Name or Material | 2,5-Di-tert-butyl-4-methoxyphenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-873-0
TSCA : No
Recommended Storage : Ambient temperatures