Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Di-tert-butyl-p-benzoquinone 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2460-77-7
C14H20O2
220.312
MFCD00019442
ZZYASVWWDLJXIM-UHFFFAOYSA-N
2,5-di-tert-butyl-1,4-benzoquinone,2,5-di-tert-butylquinone,2,5-di-tert-butylsemiquinone,2,5-di-tert-butylbenzoquinone,2,5-di-tert-butylcyclohexa-2,5-diene-1,4-dione,2,5-di-tert-butyl-p-quinone,2,5-di-tert-butyl-p-benzoquinone,unii-g893osv02d,p-benzoquinone, 2,5-di-tert-butyl,2,5-di-t-butyl-p-benzoquinone
17161
2,5-ditert-butylcyclohexa-2,5-diene-1,4-dione
CC(C)(C)C1=CC(=O)C(=CC1=O)C(C)(C)C
2460-77-7
C14H20O2
220.312
MFCD00019442
ZZYASVWWDLJXIM-UHFFFAOYSA-N
2,5-di-tert-butyl-1,4-benzoquinone,2,5-di-tert-butylquinone,2,5-di-tert-butylsemiquinone,2,5-di-tert-butylbenzoquinone,2,5-di-tert-butylcyclohexa-2,5-diene-1,4-dione,2,5-di-tert-butyl-p-quinone,2,5-di-tert-butyl-p-benzoquinone,unii-g893osv02d,p-benzoquinone, 2,5-di-tert-butyl,2,5-di-t-butyl-p-benzoquinone
17161
2,5-ditert-butylcyclohexa-2,5-diene-1,4-dione
CC(C)(C)C1=CC(=O)C(=CC1=O)C(C)(C)C
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color Yellow
Melting Point 152°C to 154°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula [(CH3)3C]2C6H2(=O)2
Formula Weight 220.31
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2, 5-Di-tert-butyl-p-benzoquinone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.