Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Di-tert-butylhydroquinone, 99%, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
Packaging:
Plastic bottle
88-58-4
C14H22O2
222.33
MFCD00008825
JZODKRWQWUWGCD-UHFFFAOYSA-N
2,5-di-tert-butylhydroquinone,2,5-di-tert-butylbenzene-1,4-diol,dibug,dtbhq,dybug,santovar o,di-t-butylhydroquinone,2,5-di-tert-butylquinol,nonflex alba,2,5-di-t-butylhydroquinone
2374
CHEBI:41094
2,5-ditert-butylbenzene-1,4-diol
CC(C)(C)C1=CC(=C(C=C1O)C(C)(C)C)O
88-58-4
C14H22O2
222.33
MFCD00008825
JZODKRWQWUWGCD-UHFFFAOYSA-N
2,5-di-tert-butylhydroquinone,2,5-di-tert-butylbenzene-1,4-diol,dibug,dtbhq,dybug,santovar o,di-t-butylhydroquinone,2,5-di-tert-butylquinol,nonflex alba,2,5-di-t-butylhydroquinone
2374
CHEBI:41094
2,5-ditert-butylbenzene-1,4-diol
CC(C)(C)C1=CC(=C(C=C1O)C(C)(C)C)O
Flash Point 216°C
Packaging Plastic bottle
Infrared Spectrum Authentic
Color Beige to Beige-Pink
Melting Point 212°C to 218°C
Density 216g/mL
Quantity 2.5kg
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in aqueous alkali,soluble in acetone,alcohol and benzene,practically insoluble in toluene and hexane
Formula Weight 222.33
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2, 5-Di-tert-butylhydroquinone
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.