Alfa Aesar™ 2,5-Diacetoxytoluene, 97%

Manufacturer: Alfa Aesar™ B2315622

Quantity 100g

 View more versions of this product

Catalog No. AAB2315622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 717-27-1
    Molecular Formula C11H12O4
    Formula Weight 208.22
    MDL Number MFCD00061098
    Beilstein 1967373
    Synonym Methylhydroquinone diacetate
    Chemical Name or Material 2,5-Diacetoxytoluene

    Specifications

    Formula Weight 208.22
    Molecular Formula C11H12O4
    MDL Number MFCD00061098
    CAS 717-27-1
    Synonym Methylhydroquinone diacetate
    Melting Point 40° to 42°C
    Quantity 100g
    Beilstein 1967373
    Chemical Name or Material 2,5-Diacetoxytoluene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit