Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride, 98%, Acros Organics

Click to view available options
Quantity:
5g
56830-58-1
C4H7ClN4O2
178.576
MFCD00082893
IIVYOQGIZSPVFZ-UHFFFAOYSA-N
2,5-diamino-4,6-dihydroxypyrimidine hydrochloride,2,5-diaminopyrimidine-4,6-diol hydrochloride,2,5-diamino-4,6-dihydroxypyrimidinehydrochloride,2,5-diamino-6-hydroxy-3h-pyrimidin-4-one hydrochloride,diaminopyrimidine-4,6-diol hydrochloride,2,5-diamino-4,6-dihydroxypyrimidine hcl,2,5-diamino-4-hydroxy-1h-pyrimidin-6-one hydrochloride,4 1h-pyrimidinone, 2,5-diamino-6-hydroxy-, monohydrochloride,2,5-diaminopyrimidine-4,6-diol, chloride,2,5-diamino-6-hydroxy-3h-pyrimidin-4-one; hydrochloride
2733980
2,5-diamino-4-hydroxy-1H-pyrimidin-6-one;hydrochloride
C1(=C(N=C(NC1=O)N)O)N.Cl
56830-58-1
C4H7ClN4O2
178.576
MFCD00082893
IIVYOQGIZSPVFZ-UHFFFAOYSA-N
2,5-diamino-4,6-dihydroxypyrimidine hydrochloride,2,5-diaminopyrimidine-4,6-diol hydrochloride,2,5-diamino-4,6-dihydroxypyrimidinehydrochloride,2,5-diamino-6-hydroxy-3h-pyrimidin-4-one hydrochloride,diaminopyrimidine-4,6-diol hydrochloride,2,5-diamino-4,6-dihydroxypyrimidine hcl,2,5-diamino-4-hydroxy-1h-pyrimidin-6-one hydrochloride,4 1h-pyrimidinone, 2,5-diamino-6-hydroxy-, monohydrochloride,2,5-diaminopyrimidine-4,6-diol, chloride,2,5-diamino-6-hydroxy-3h-pyrimidin-4-one; hydrochloride
2733980
2,5-diamino-4-hydroxy-1H-pyrimidin-6-one;hydrochloride
C1(=C(N=C(NC1=O)N)O)N.Cl
Quantity 5g
Color Orange-Pink
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 178.58
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Diamino-4, 6-dihydroxypyrimidine hydrochloride, 98%