Alfa Aesar™ 2,5-Dibromo-3-n-decylthiophene, 97%

Manufacturer: Alfa Aesar™ H55497MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55497MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 158956-23-1
    Formula Weight 382.2
    MDL Number MFCD00274314
    Chemical Name or Material 2,5-Dibromo-3-n-decylthiophene

    Specifications

    Formula Weight 382.2
    Boiling Point 168° to 170°C (2mmHg)
    Physical Form Liquid
    CAS 158956-23-1
    Refractive Index 1.5380
    Density 1.372g/mL
    Quantity 250mg
    MDL Number MFCD00274314
    Chemical Name or Material 2,5-Dibromo-3-n-decylthiophene
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit