Alfa Aesar™ 2,5-Dibromo-3-n-dodecylthiophene, 97%

Manufacturer: Alfa Aesar™ H5595806

Quantity 5g

 View more versions of this product

Catalog No. AAH5595806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 148256-63-7
    Molecular Formula C16H26Br2S
    Formula Weight 410.25
    MDL Number MFCD00274315
    Chemical Name or Material 2,5-Dibromo-3-n-dodecylthiophene

    Specifications

    Formula Weight 410.25
    Flash Point >113°C (235°F)
    Boiling Point 135°C (2.5mmHg)
    Quantity 5g
    Molecular Formula C16H26Br2S
    MDL Number MFCD00274315
    CAS 148256-63-7
    Chemical Name or Material 2,5-Dibromo-3-n-dodecylthiophene
    Assay 97%

    Description

    Applications
    2,5-Dibromo-3-n-dodecylthiophene is used as an OLED intermediate, flavor and fragrance intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit