Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-3-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
3430-18-0
C6H5Br2N
250.921
MFCD02093085
LIMXEVCFAUTBCK-UHFFFAOYSA-N
2,5-dibromo-3-picoline,2,5-dibromo-3-methyl-pyridine,pyridine, 2,5-dibromo-3-methyl,2,5-dibromo-3-methylpyridine,2,5-dibrom-3-methylpyridin,pubchem1138,acmc-209i6e,ksc495g9h,2,5-dibromo-3-methyl pyridine
817680
2,5-dibromo-3-methylpyridine
CC1=CC(=CN=C1Br)Br
3430-18-0
C6H5Br2N
250.921
MFCD02093085
LIMXEVCFAUTBCK-UHFFFAOYSA-N
2,5-dibromo-3-picoline,2,5-dibromo-3-methyl-pyridine,pyridine, 2,5-dibromo-3-methyl,2,5-dibromo-3-methylpyridine,2,5-dibrom-3-methylpyridin,pubchem1138,acmc-209i6e,ksc495g9h,2,5-dibromo-3-methyl pyridine
817680
2,5-dibromo-3-methylpyridine
CC1=CC(=CN=C1Br)Br
Color White to Beige
Quantity 1g
Melting Point 42°C to 47°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 20, V,5, 538
Formula Weight 251
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Dibromo-3-picoline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning