Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dibromo-4-fluorotoluene, 96%

Click to view available options
Quantity:
25g
100g
500g
134332-29-9
C7H5Br2F
267.923
MFCD00044738
CAJWWHAZRJAYKR-UHFFFAOYSA-N
2,5-dibromo-4-fluorotoluene,2,5-dibromo-1-fluoro-4-methylbenzene
2723682
1,4-dibromo-2-fluoro-5-methylbenzene
CC1=CC(=C(C=C1Br)F)Br
134332-29-9
C7H5Br2F
267.923
MFCD00044738
CAJWWHAZRJAYKR-UHFFFAOYSA-N
2,5-dibromo-4-fluorotoluene,2,5-dibromo-1-fluoro-4-methylbenzene
2723682
1,4-dibromo-2-fluoro-5-methylbenzene
CC1=CC(=C(C=C1Br)F)Br
Boiling Point 92°C to 94°C (1mmHg)
Quantity 100g
Formula Weight 267.92
Percent Purity 96%
Chemical Name or Material 2,5-Dibromo-4-fluorotoluene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning