Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-4-fluorotoluene 96%, Acros Organics

Click to view available options
Quantity:
100g
134332-29-9
C7H5Br2F
267.923
MFCD00044738
CAJWWHAZRJAYKR-UHFFFAOYSA-N
2,5-dibromo-4-fluorotoluene,2,5-dibromo-1-fluoro-4-methylbenzene
2723682
1,4-dibromo-2-fluoro-5-methylbenzene
CC1=CC(=C(C=C1Br)F)Br
134332-29-9
C7H5Br2F
267.923
MFCD00044738
CAJWWHAZRJAYKR-UHFFFAOYSA-N
2,5-dibromo-4-fluorotoluene,2,5-dibromo-1-fluoro-4-methylbenzene
2723682
1,4-dibromo-2-fluoro-5-methylbenzene
CC1=CC(=C(C=C1Br)F)Br
Quantity 100g
Infrared Spectrum Authentic
Color Colorless to Yellow
Refractive Index 1.5805 to 1.5825 (20°C, 589nm)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula (Br)2C6H2(CH3)F
Formula Weight 267.91
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 2, 5-Dibromo-4-fluorotoluene, 96%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA