Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dibromo-4-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
3430-26-0
C6H5Br2N
250.921
MFCD00234955
WWJLJUAHQHXDGM-UHFFFAOYSA-N
2,5-dibromo-4-picoline,pyridine, 2,5-dibromo-4-methyl,2,5-dibromo-4-methyl-pyridine,pubchem2411,2 pound not5-dibromo-4-methylpyridine,acmc-209i6i,2, 5-dibromo-4-picoline,2,5-dibromo-4-methypyridine,ksc495g9l,tpc-py104
2734429
2,5-dibromo-4-methylpyridine
CC1=CC(=NC=C1Br)Br
3430-26-0
C6H5Br2N
250.921
MFCD00234955
WWJLJUAHQHXDGM-UHFFFAOYSA-N
2,5-dibromo-4-picoline,pyridine, 2,5-dibromo-4-methyl,2,5-dibromo-4-methyl-pyridine,pubchem2411,2 pound not5-dibromo-4-methylpyridine,acmc-209i6i,2, 5-dibromo-4-picoline,2,5-dibromo-4-methypyridine,ksc495g9l,tpc-py104
2734429
2,5-dibromo-4-methylpyridine
CC1=CC(=NC=C1Br)Br
Infrared Spectrum Authentic
Color Orange to Yellow
Melting Point 37.0°C to 41.0°C
Quantity 1g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 251
Physical Form Low Melting Solid
Percent Purity 98%
Chemical Name or Material 2, 5-Dibromo-4-picoline
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning