Alfa Aesar™ 2,5-Dibromobenzaldehyde, 97%

Manufacturer: Alfa Aesar™ H3168914

Quantity 25g

 View more versions of this product

Catalog No. AAH3168914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74553-29-0
    Molecular Formula C7H4Br2O
    Formula Weight 263.93
    MDL Number MFCD00462864
    Chemical Name or Material 2,5-Dibromobenzaldehyde

    Specifications

    Formula Weight 263.93
    Melting Point 88° to 102°C
    Quantity 25g
    Molecular Formula C7H4Br2O
    MDL Number MFCD00462864
    CAS 74553-29-0
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2,5-Dibromobenzaldehyde
    Assay 97%

    Description

    Applications
    2,5-Dibromobenzaldehyde is used as intermediate, it is used to produce other chemicals, it can react with Ethane-1,2-diol to get 2-(2,5-Dibromo-phenyl)-[1,3]dioxolane. The reaction occurs with reagent p-toluenesulfonic acid, solvent toluene.

    Solubility
    Soluble in water.

    Notes
    Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit