Alfa Aesar™ 2,5-Dichloro-1,4-benzoquinone, 98%

Manufacturer: Alfa Aesar™ H2715209

Quantity 10g

 View more versions of this product

Catalog No. AAH2715209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 615-93-0
    Molecular Formula C6H2Cl2O2
    Formula Weight 176.98
    MDL Number MFCD00041736
    Chemical Name or Material 2,5-Dichloro-1,4-benzoquinone

    Specifications

    Formula Weight 176.98
    TSCA TSCA
    Molecular Formula C6H2Cl2O2
    MDL Number MFCD00041736
    CAS 615-93-0
    Melting Point 158° to 162°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DK3990000
    Quantity 10g
    Chemical Name or Material 2,5-Dichloro-1,4-benzoquinone
    Assay 98%

    Description

    Applications
    2,5-Dichloro-1,4-benzoquinone, is used as a agrochemical, pharmaceutical intermediate. It is also used in the organic chemical synthesis intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit