Alfa Aesar™ 2,5-Dichloro-3-methylthiophene, 96%

Manufacturer: Alfa Aesar™ H2712903

Quantity 1g

 View more versions of this product

Catalog No. AAH2712903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17249-90-0
    Molecular Formula C5H4Cl2S
    Formula Weight 167.06
    MDL Number MFCD00022490
    UN Number UN2810
    Chemical Name or Material 2,5-Dichloro-3-methylthiophene

    Specifications

    Formula Weight 167.06
    CAS 17249-90-0
    Flash Point 77°C (171°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H227-H302-H312-H315-H319
    Toxic if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C5H4Cl2S
    MDL Number MFCD00022490
    UN Number UN2810
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 2,5-Dichloro-3-methylthiophene
    Assay 96%

    GHS H Statement
    H331-H227-H302-H312-H315-H319
    Toxic if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit