Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloro-3-nitropyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
21427-62-3
C5H2Cl2N2O2
192.99
MFCD06658963
OBUGJYJQJWMOQO-UHFFFAOYSA-N
2,5-dichloro-3-nitro-pyridine,pyridine, 2,5-dichloro-3-nitro,2.5-dichloro-3-nitropyridine,2,5-dichloro-3-nitro pyridine,pubchem2267,acmc-1cdlk,2,5dichloro-3-nitropyridine,ksc495e7t,2,5 dichloro-3-nitropyridine,2,5 dichloro-3-nitro pyridine
8063331
2,5-dichloro-3-nitropyridine
C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]
21427-62-3
C5H2Cl2N2O2
192.99
MFCD06658963
OBUGJYJQJWMOQO-UHFFFAOYSA-N
2,5-dichloro-3-nitro-pyridine,pyridine, 2,5-dichloro-3-nitro,2.5-dichloro-3-nitropyridine,2,5-dichloro-3-nitro pyridine,pubchem2267,acmc-1cdlk,2,5dichloro-3-nitropyridine,ksc495e7t,2,5 dichloro-3-nitropyridine,2,5 dichloro-3-nitro pyridine
8063331
2,5-dichloro-3-nitropyridine
C1=C(C(=NC=C1Cl)Cl)[N+](=O)[O-]
Melting Point 39°C to 45°C
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige-Green to Orange
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Beilstein 20, IV, 2532
Formula Weight 192.99
Physical Form Crystalline Powder and/or Chunks
Percent Purity 97%
Chemical Name or Material 2, 5-Dichloro-3-nitropyridine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning