Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloro-4-phenylenediamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
20103-09-7
C6H6Cl2N2
177.028
QAYVHDDEMLNVMO-UHFFFAOYSA-N
2,5-dichloro-1,4-phenylenediamine, 2,5-dichloro-p-phenylenediamine, 2,5-dichloro-1,4-benzenediamine, 1,4-diamino-2,5-dichlorobenzene, 1,4-benzenediamine, 2,5-dichloro, unii-9ccw77baxp, 1,4-diamino-3,6-dichlorobenzene, p-phenylenediamine, 2,5-dichloro, 9ccw77baxp, benzene, 2,5-dichloro, 1,4-diamino
29949
2,5-dichlorobenzene-1,4-diamine
C1=C(C(=CC(=C1Cl)N)Cl)N
20103-09-7
C6H6Cl2N2
177.028
QAYVHDDEMLNVMO-UHFFFAOYSA-N
2,5-dichloro-1,4-phenylenediamine, 2,5-dichloro-p-phenylenediamine, 2,5-dichloro-1,4-benzenediamine, 1,4-diamino-2,5-dichlorobenzene, 1,4-benzenediamine, 2,5-dichloro, unii-9ccw77baxp, 1,4-diamino-3,6-dichlorobenzene, p-phenylenediamine, 2,5-dichloro, 9ccw77baxp, benzene, 2,5-dichloro, 1,4-diamino
29949
2,5-dichlorobenzene-1,4-diamine
C1=C(C(=CC(=C1Cl)N)Cl)N
Melting Point 164°C to 168°C
Infrared Spectrum Authentic
Color Brown to Gray-Pink
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl2C6H2(NH2)2
Beilstein 13, 118
Formula Weight 177.03
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Dichloro-4-phenylenediamine
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 243-512-1

RTECSNumber : SS9150000

TSCA : TSCA