Alfa Aesar™ 2,5-Dichloro-4-(trifluoromethyl)pyridine, 97%

Manufacturer: Alfa Aesar™ H3344203

Quantity 1g

 View more versions of this product

Catalog No. AAH3344203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89719-92-6
    Molecular Formula C6H2Cl2F3N
    Formula Weight 215.99
    MDL Number MFCD09749663
    UN Number UN1993
    Chemical Name or Material 2,5-Dichloro-4-(trifluoromethyl)pyridine

    Specifications

    Formula Weight 215.99
    Molecular Formula C6H2Cl2F3N
    MDL Number MFCD09749663
    CAS 89719-92-6
    UN Number UN1993
    Boiling Point 76°C (18mmHg)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H226-H315-H319-H335
    Toxic if swallowed.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Quantity 1g
    Chemical Name or Material 2,5-Dichloro-4-(trifluoromethyl)pyridine
    Assay 97%

    Description

    Applications
    2-chloro-6-(trifluoromethyl)pyridine, 2-chloro-5-(trifluoromethyl)pyridine and 3-chloro-4-(trifluoromethyl)pyridine were each converted into the three possible carboxylic acids 2, 4, 6,8, 10, 12, 16, 17 and 20. This was achieved by employing several, but not all of the organometallic “toolbox methods”: transformation of a more basic organometallic species into a less basic isomer by transmetalation-equilibration.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, aluminum. Protection under nitrogen and lower temperature is recommended(below 4 deg C).

    GHS H Statement
    H301-H226-H315-H319-H335
    Toxic if swallowed.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit