Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dichloroacetophenone, 96%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
100g
Packaging:
Glass bottle
2476-37-1
C8H6Cl2O
189.04
MFCD00000607
CYNFEPKQDJHIMV-UHFFFAOYSA-N
2',5'-dichloroacetophenone,1-2,5-dichlorophenyl ethanone,2,5-dichloroacetophenone,ethanone, 1-2,5-dichlorophenyl,acetophenone, 2',5'-dichloro,1-2,5-dichlorophenyl ethan-1-one,1-acetyl-2,5-dichlorobenzene,2,5-dichloracetophenon,pubchem10586,acmc-1cmfm
75587
1-(2,5-dichlorophenyl)ethanone
CC(=O)C1=C(C=CC(=C1)Cl)Cl
2476-37-1
C8H6Cl2O
189.04
MFCD00000607
CYNFEPKQDJHIMV-UHFFFAOYSA-N
2',5'-dichloroacetophenone,1-2,5-dichlorophenyl ethanone,2,5-dichloroacetophenone,ethanone, 1-2,5-dichlorophenyl,acetophenone, 2',5'-dichloro,1-2,5-dichlorophenyl ethan-1-one,1-acetyl-2,5-dichlorobenzene,2,5-dichloracetophenon,pubchem10586,acmc-1cmfm
75587
1-(2,5-dichlorophenyl)ethanone
CC(=O)C1=C(C=CC(=C1)Cl)Cl
Melting Point 11.0°C to 13.0°C
Density 1.3120g/mL
Boiling Point 118.0°C (12.0 mmHg)
Quantity 50g
Infrared Spectrum Authentic
Color Brown to Yellow
Refractive Index 1.5604 to 1.5624
Specific Gravity 1.312
Flash Point >110°C
Packaging Glass bottle
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula Cl2C6H3COCH3
Formula Weight 189.04
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 2', 5'-Dichloroacetophenone, 96%
Show More Show Less

EINECSNumber : 219-605-8

TSCA : TSCA