Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dichlorobenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
135145-90-3
C6H5BCl2O2
190.814
MFCD01863182
NNTFPBXQPOQRBT-UHFFFAOYSA-N
2,5-dichlorobenzeneboronic acid,2,5-dichlorophenyl boronic acid,2,5-dichlorophenylboronicacid,2,5-dichlorophenyl boranediol,boronic acid, 2,5-dichlorophenyl,pubchem1811,acmc-209bxy,ksc174k3h,2,5-dichloro-phenylboronic acid
2773371
(2,5-dichlorophenyl)boronic acid
B(C1=C(C=CC(=C1)Cl)Cl)(O)O
135145-90-3
C6H5BCl2O2
190.814
MFCD01863182
NNTFPBXQPOQRBT-UHFFFAOYSA-N
2,5-dichlorobenzeneboronic acid,2,5-dichlorophenyl boronic acid,2,5-dichlorophenylboronicacid,2,5-dichlorophenyl boranediol,boronic acid, 2,5-dichlorophenyl,pubchem1811,acmc-209bxy,ksc174k3h,2,5-dichloro-phenylboronic acid
2773371
(2,5-dichlorophenyl)boronic acid
B(C1=C(C=CC(=C1)Cl)Cl)(O)O
Quantity 5g
Melting Point ~210°C to 216°C
Beilstein 4801290
Formula Weight 190.82
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning