Alfa Aesar™ 2,5-Dichlorobenzenesulfonic acid dihydrate, 97%

Manufacturer: Alfa Aesar™ B2565022

Quantity 100g

 View more versions of this product

Catalog No. AAB2565022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38484-94-5
    Molecular Formula C6H4Cl2O3S·2H2O
    Formula Weight 263.09 (227.07 Anhydrous)
    MDL Number MFCD08271971
    UN Number UN2585
    Chemical Name or Material 2,5-Dichlorobenzenesulfonic acid dihydrate

    Specifications

    Formula Weight 263.09 (227.07 Anhydrous)
    Quantity 100g
    TSCA TSCA
    Molecular Formula C6H4Cl2O3S·2H2O
    MDL Number MFCD08271971
    UN Number UN2585
    Melting Point 98° to 102°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
    CAS 38484-94-5
    Chemical Name or Material 2,5-Dichlorobenzenesulfonic acid dihydrate
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit