Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorobenzenesulfonyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
5402-73-3
C6H3Cl3O2S
245.498
MFCD00007429
BXCOSWRSIISQSL-UHFFFAOYSA-N
2,5-dichlorobenzene-1-sulfonyl chloride, 2,5-dichlorobenzenesulphonyl chloride, 2,5-dichlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2,5-dichloro, 2,5-dichloro-benzenesulfonyl chloride, 2,5-dichlorophenylsulfonyl chloride, 2,5-dichlorophenyl chlorosulfone, pubchem5116, acmc-209lcc, bxcoswrsiisqsl-uhfffaoysa
79401
2,5-dichlorobenzenesulfonyl chloride
C1=CC(=C(C=C1Cl)S(=O)(=O)Cl)Cl
5402-73-3
C6H3Cl3O2S
245.498
MFCD00007429
BXCOSWRSIISQSL-UHFFFAOYSA-N
2,5-dichlorobenzene-1-sulfonyl chloride, 2,5-dichlorobenzenesulphonyl chloride, 2,5-dichlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2,5-dichloro, 2,5-dichloro-benzenesulfonyl chloride, 2,5-dichlorophenylsulfonyl chloride, 2,5-dichlorophenyl chlorosulfone, pubchem5116, acmc-209lcc, bxcoswrsiisqsl-uhfffaoysa
79401
2,5-dichlorobenzenesulfonyl chloride
C1=CC(=C(C=C1Cl)S(=O)(=O)Cl)Cl
Infrared Spectrum Authentic
Color Beige
Melting Point 35°C to 39°C
Packaging Glass bottle
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl2C6H3SO2Cl
Beilstein 11, I, 15
Formula Weight 245.51
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Dichlorobenzenesulfonyl chloride, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 226-453-6

TSCA : TSCA