Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorobenzyl bromide, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
85482-13-9
C7H5BrCl2
239.93
UUVDOPTUDWJHFK-UHFFFAOYSA-N
2,5-dichlorobenzyl bromide,2-bromomethyl-1,4-dichlorobenzene,2,5-dichlorobenzylbromide,1-bromomethyl-2,5-dichlorobenzene,alpha-bromo-2,5-dichlorotoluene,benzene, 2-bromomethyl-1,4-dichloro,2-bromomethyl-1,4-dichloro-benzene,pubchem10208,acmc-1bjf0,2,5-di-chloro-benzyl bromide
3495744
2-(bromomethyl)-1,4-dichlorobenzene
C1=CC(=C(C=C1Cl)CBr)Cl
85482-13-9
C7H5BrCl2
239.93
UUVDOPTUDWJHFK-UHFFFAOYSA-N
2,5-dichlorobenzyl bromide,2-bromomethyl-1,4-dichlorobenzene,2,5-dichlorobenzylbromide,1-bromomethyl-2,5-dichlorobenzene,alpha-bromo-2,5-dichlorotoluene,benzene, 2-bromomethyl-1,4-dichloro,2-bromomethyl-1,4-dichloro-benzene,pubchem10208,acmc-1bjf0,2,5-di-chloro-benzyl bromide
3495744
2-(bromomethyl)-1,4-dichlorobenzene
C1=CC(=C(C=C1Cl)CBr)Cl
Infrared Spectrum Authentic
Melting Point 41.0°C to 43.0°C
Boiling Point 134.0°C to 138.0°C (14.0 Torr)
Quantity 25g
Assay Percent Range 96% min. (GC)
Formula Weight 239.93
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzyl bromide

GHS H Statement Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger