Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dichloronicotinic acid, 98%

Click to view available options
Quantity:
1g
5g
59782-85-3
C6H3Cl2NO2
191.995
MFCD01861979
SXQSMLIMBNMUNB-UHFFFAOYSA-N
2,5-dichloronicotinic acid,2,5-dichloronicotinicacid,3-pyridinecarboxylic acid, 2,5-dichloro,2,5-dichloro-3-pyridinecarboxylic acid,pubchem2601,acmc-209mfn,pyni16,2,5-dichloro-nicotinic acid,2,5-dichloronicotinic acid?,ksc269g2r
2736085
2,5-dichloropyridine-3-carboxylic acid
C1=C(C(=NC=C1Cl)Cl)C(=O)O
59782-85-3
C6H3Cl2NO2
191.995
MFCD01861979
SXQSMLIMBNMUNB-UHFFFAOYSA-N
2,5-dichloronicotinic acid,2,5-dichloronicotinicacid,3-pyridinecarboxylic acid, 2,5-dichloro,2,5-dichloro-3-pyridinecarboxylic acid,pubchem2601,acmc-209mfn,pyni16,2,5-dichloro-nicotinic acid,2,5-dichloronicotinic acid?,ksc269g2r
2736085
2,5-dichloropyridine-3-carboxylic acid
C1=C(C(=NC=C1Cl)Cl)C(=O)O
Quantity 1g
Density 1.61g/mL
Color White
Melting Point 156°C to 158°C
Beilstein 389586
Formula Weight 192
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2,5-Dichloronicotinic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning