Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dichlorophenol, 98%

Click to view available options
Quantity:
25g
100g
500g
583-78-8
C6H4Cl2O
162.997
MFCD00002174
RANCECPPZPIPNO-UHFFFAOYSA-N
phenol, 2,5-dichloro,2,5-dichloro-phenol,2,5-dcp,unii-3b11g9akba,3,6-dichlorophenol,ccris 5903,3b11g9akba,dsstox_cid_5003,dsstox_rid_77621,dsstox_gsid_25003
66
CHEBI:27929
2,5-dichlorophenol
C1=CC(=C(C=C1Cl)O)Cl
583-78-8
C6H4Cl2O
162.997
MFCD00002174
RANCECPPZPIPNO-UHFFFAOYSA-N
phenol, 2,5-dichloro,2,5-dichloro-phenol,2,5-dcp,unii-3b11g9akba,3,6-dichlorophenol,ccris 5903,3b11g9akba,dsstox_cid_5003,dsstox_rid_77621,dsstox_gsid_25003
66
CHEBI:27929
2,5-dichlorophenol
C1=CC(=C(C=C1Cl)O)Cl
Quantity 25g
Boiling Point 211°C
Melting Point 55°C to 58°C
Odor Phenol-like
UN Number UN2020
Beilstein 1907692
Formula Weight 163
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorophenol

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID

EINECSNumber : 209-520-4

RTECSNumber : SK8600000

TSCA : Yes

Recommended Storage : Ambient temperatures