Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,5-Dichlorophenol, 98%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Boiling Point | 211°C |
Quantity | 500g |
Melting Point | 55°C to 58°C |
Odor | Phenol-like |
UN Number | UN2020 |
Beilstein | 1907692 |
Formula Weight | 163 |
Percent Purity | 98% |
Chemical Name or Material | 2,5-Dichlorophenol |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber : 209-520-4
RTECSNumber : SK8600000
TSCA : Yes
Recommended Storage : Ambient temperatures