Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dichloropyridine, 98%

Click to view available options
Quantity:
25g
50g
250g
16110-09-1
C5H3Cl2N
147.986
MFCD00006239
GCTFDMFLLBCLPF-UHFFFAOYSA-N
pyridine, 2,5-dichloro,2,5-dichloro-pyridine,2,5-dichloro pyridine,ccris 1718,2,5-dichlorpyridin,2,5-dichloropyridin,2,5-dichioropyridine,2.5-dichloropyridine,pubchem1199,2,5 dichloropyridine
27685
2,5-dichloropyridine
C1=CC(=NC=C1Cl)Cl
16110-09-1
C5H3Cl2N
147.986
MFCD00006239
GCTFDMFLLBCLPF-UHFFFAOYSA-N
pyridine, 2,5-dichloro,2,5-dichloro-pyridine,2,5-dichloro pyridine,ccris 1718,2,5-dichlorpyridin,2,5-dichloropyridin,2,5-dichioropyridine,2.5-dichloropyridine,pubchem1199,2,5 dichloropyridine
27685
2,5-dichloropyridine
C1=CC(=NC=C1Cl)Cl
Applications
2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.

Solubility
Insoluble in water.

Notes
Incompatible with Strong acids, strong bases, strong oxidizing agents.
Melting Point 59°C to 62°C
Flash Point 112°C (233°F)
Quantity 250g
Odor Unpleasant
Boiling Point 190°C to 191°C
UN Number UN2811
Beilstein 108886
Solubility Information Insoluble in water.
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 2,5-Dichloropyridine
Show More Show Less

GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 240-278-2

RTECSNumber : US8225000

TSCA : No

Recommended Storage : Ambient temperatures