Alfa Aesar™ 2,5-Dichlorothiobenzamide, 97%

Manufacturer: Alfa Aesar™ H3232303

Quantity 1g

 View more versions of this product

Catalog No. AAH3232303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69622-81-7
    Molecular Formula C7H5Cl2NS
    Formula Weight 206.09
    MDL Number MFCD09025686
    UN Number UN2811
    Chemical Name or Material 2,5-Dichlorothiobenzamide

    Specifications

    Formula Weight 206.09
    Molecular Formula C7H5Cl2NS
    UN Number UN2811
    Melting Point 88° to 92°C
    MDL Number MFCD09025686
    CAS 69622-81-7
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 2,5-Dichlorothiobenzamide
    Assay 97%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit