Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Difluoro-4-hydroxybenzaldehyde, 99%

Manufacturer:  Alfa Aesar™ H3152806

Quantity 5g

 View more versions of this product

Catalog No. AAH3152806


  Check Availability
Add to cart

Specifications

Description

Applications
2,5-Difluoro-4-hydroxybenzaldehyde is used as primary and secondary intermediate and as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.
Description & Specifications

Chemical Identifiers

918523-99-6
158.11
2,5-Difluoro-4-hydroxybenzaldehyde
C7H4F2O2
MFCD04112521

Specifications

145° to 148°C
918523-99-6
158.11
2,5-Difluoro-4-hydroxybenzaldehyde
5g
C7H4F2O2
MFCD04112521
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit