Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluoroaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
25mL
Packaging:
Glass bottle
367-30-6
C6H5F2N
129.11
MFCD00007651
YNOOQIUSYGWMSS-UHFFFAOYSA-N
benzenamine, 2,5-difluoro,2,5-difluorobenzenamine,2,5-difluoro-phenylamine,aniline, 2,5-difluoro,2.5-difluoroaniline,aniline,5-difluoro,pubchem1531,2,5-difluoro-aniline,2,5-difluorophenylamine,benzenamine,5-difluoro
67775
2,5-difluoroaniline
C1=CC(=C(C=C1F)N)F
367-30-6
C6H5F2N
129.11
MFCD00007651
YNOOQIUSYGWMSS-UHFFFAOYSA-N
benzenamine, 2,5-difluoro,2,5-difluorobenzenamine,2,5-difluoro-phenylamine,aniline, 2,5-difluoro,2.5-difluoroaniline,aniline,5-difluoro,pubchem1531,2,5-difluoro-aniline,2,5-difluorophenylamine,benzenamine,5-difluoro
67775
2,5-difluoroaniline
C1=CC(=C(C=C1F)N)F
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.2880g/mL
Color Brown
Quantity 25mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula F2C6H3NH2
Formula Weight 129.11
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2, 5-Difluoroaniline, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-690-1

TSCA : TSCA

Recommended Storage : Product darkens on storage