Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Difluorobenzeneboronic acid, 96%

Click to view available options
Quantity:
1g
5g
25g
193353-34-3
C6H5BF2O2
157.911
MFCD01863171
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2,5-difluorophenyl boronic acid,2,5-difluorobenzeneboronic acid,2,5-difluorophenyl boranediol,boronic acid, 2,5-difluorophenyl,2,5-difluorophenylboronicacid,2,5-fluorobenzene boronic acid,pubchem1820,acmc-1c1am,ksc174i6r,2,5-difluoro phenylboronic acid
2734335
(2,5-difluorophenyl)boronic acid
B(C1=C(C=CC(=C1)F)F)(O)O
193353-34-3
C6H5BF2O2
157.911
MFCD01863171
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2,5-difluorophenyl boronic acid,2,5-difluorobenzeneboronic acid,2,5-difluorophenyl boranediol,boronic acid, 2,5-difluorophenyl,2,5-difluorophenylboronicacid,2,5-fluorobenzene boronic acid,pubchem1820,acmc-1c1am,ksc174i6r,2,5-difluoro phenylboronic acid
2734335
(2,5-difluorophenyl)boronic acid
B(C1=C(C=CC(=C1)F)F)(O)O
Quantity 5g
Beilstein 8833254
Formula Weight 157.91
Percent Purity 96%
Chemical Name or Material 2,5-Difluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning