Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Difluorobenzeneboronic acid, 96%

Click to view available options
Quantity:
1g
5g
25g
193353-34-3
C6H5BF2O2
157.911
MFCD01863171
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2,5-difluorophenyl boronic acid,2,5-difluorobenzeneboronic acid,2,5-difluorophenyl boranediol,boronic acid, 2,5-difluorophenyl,2,5-difluorophenylboronicacid,2,5-fluorobenzene boronic acid,pubchem1820,acmc-1c1am,ksc174i6r,2,5-difluoro phenylboronic acid
2734335
(2,5-difluorophenyl)boronic acid
B(C1=C(C=CC(=C1)F)F)(O)O
193353-34-3
C6H5BF2O2
157.911
MFCD01863171
KTOJGSDLJNUAEP-UHFFFAOYSA-N
2,5-difluorophenyl boronic acid,2,5-difluorobenzeneboronic acid,2,5-difluorophenyl boranediol,boronic acid, 2,5-difluorophenyl,2,5-difluorophenylboronicacid,2,5-fluorobenzene boronic acid,pubchem1820,acmc-1c1am,ksc174i6r,2,5-difluoro phenylboronic acid
2734335
(2,5-difluorophenyl)boronic acid
B(C1=C(C=CC(=C1)F)F)(O)O
Quantity 25g
Beilstein 8833254
Formula Weight 157.91
Percent Purity 96%
Chemical Name or Material 2,5-Difluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures