Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,5-Difluorobenzoic acid, 98+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,5-Difluorobenzoic acid is used in the preparation of hydrazone derivatives, which acts as a potential antibacterial agent.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
2,5-Difluorobenzoic acid is used in the preparation of hydrazone derivatives, which acts as a potential antibacterial agent.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 127°C to 132°C |
Odor | Odorless |
Quantity | 25 g |
Beilstein | 973351 |
Solubility Information | Insoluble in water. |
Formula Weight | 158.11 |
Percent Purity | ≥98% |
Chemical Name or Material | 2,5-Difluorobenzoic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 221-060-6 |
TSCA | Yes |
storageNote1 | Incompatible with strong oxidizing agents. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction