Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzoic acid, 98+%

Used in the preparation of hydrazone derivatives. Acts as a potential antibacterial agent.

Manufacturer:  Alfa Aesar A1050303

Quantity 1g

 View more versions of this product

Catalog No. AAA1050303


  Check Availability
Add to cart

Specifications

Description

Applications
2,5-Difluorobenzoic acid is used in the preparation of hydrazone derivatives, which acts as a potential antibacterial agent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

2991-28-8
158.11
973351
C7H4F2O2
MFCD00002410
2,5-Difluorobenzoic acid

Specifications

Odorless
1g
C7H4F2O2
MFCD00002410
Insoluble in water.
≥98%
127° to 132°C
2991-28-8
158.11
973351
2,5-Difluorobenzoic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

EINECSNumber : 221-060-6

storageNote1 : Incompatible with strong oxidizing agents.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit