Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzoic acid, 98+%, Thermo Scientific™

Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
2991-28-8
C7H4F2O2
158.104
MFCD00002410
LBQMIAVIGLLBGW-UHFFFAOYSA-N
benzoic acid, 2,5-difluoro, 2,5-difluorobenzoicacid, 2,5-difluoro-benzoic acid, pubchem4935, 2,5-difluorobenzic acid, acmc-1cj4a, ksc206g1b, rarechem al bo 0019, lbqmiavigllbgw-uhfffaoysa, 2,5-difluorobenzoic acid
76339
2,5-difluorobenzoic acid
C1=CC(=C(C=C1F)C(=O)O)F
This item is not returnable. View return policy
2991-28-8
C7H4F2O2
158.104
MFCD00002410
LBQMIAVIGLLBGW-UHFFFAOYSA-N
benzoic acid, 2,5-difluoro, 2,5-difluorobenzoicacid, 2,5-difluoro-benzoic acid, pubchem4935, 2,5-difluorobenzic acid, acmc-1cj4a, ksc206g1b, rarechem al bo 0019, lbqmiavigllbgw-uhfffaoysa, 2,5-difluorobenzoic acid
76339
2,5-difluorobenzoic acid
C1=CC(=C(C=C1F)C(=O)O)F

Used in the preparation of hydrazone derivatives. Acts as a potential antibacterial agent.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,5-Difluorobenzoic acid is used in the preparation of hydrazone derivatives, which acts as a potential antibacterial agent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Melting Point 127°C to 132°C
Odor Odorless
Quantity 5 g
Beilstein 973351
Solubility Information Insoluble in water.
Formula Weight 158.11
Percent Purity ≥98%
Chemical Name or Material 2,5-Difluorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 221-060-6

TSCA : Yes

storageNote1 : Incompatible with strong oxidizing agents.